1-Heptanesulfonic acid sodium salt, hydrate ~98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium heptane-1-sulfonate, Sodium 1-heptanesulfonate, Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate

0215123925
IC15123925EA 204.74 USD
IC15123925
1-Heptanesulfonic acid sodium salt, hydrate ~98%, white powder
1-Heptanesulphonic acid sodium salt

Sodium 1-heptanesulfonate is a commonly used ion pairing reagent for HPLC, including analyses of peptides and proteins. It has also been used in high performance capillary electrophoresis analysis of peptides.


Soluble in water (10% w/v - clear, colorless solution), ethanol or methanol (0,4% w/v - clear solution).


Caution: Research Use Only (RUO).

Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
MDL Number: MFCD00007543
CAS Number: 22767-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab18d35036f2552fab9aaf0abac76b562721b6bf&vid=ab18d35036f2552fab9aaf0abac76b562721b6bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~98%
Moisture 7.4 - 9.0%
FTIR Conforms to Standard
Moisture Content 7.4-9.0%
UV/VIS:(60 mg/ml in water adjusted to pH 3) OD210 nm 1.0
OD220 nm 0.1
OD254 nm 0.04
OD280 nm 0.04

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR