Ellagic acid, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215102901
IC15102901EA 163.48 USD
IC15102901
Ellagic acid, powder
Ellagic acid

Soluble in 1 M Sodium Hydroxide (10 mg/mL - dark brown to green solution) or other alkalies; soluble in pyridine. Slightly soluble in water or ethanol.

Formula: C₁₄H₆O₈
MDL Number: MFCD00006914
CAS Number: 476-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR