Cresyl violet (acetate), dark green powder

Supplier: 0000012207

0215072701 0215072705
IC15072701EA 55.12 USD
IC15072701 IC15072705
Cresyl violet (acetate), dark green powder
Cresyl violet (acetate)

Cresyl violet acetate is used for staining neve tissue and nerve cells and fresh tumor tissue for biopsy.

Formula: C₁₈H₁₅N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00013151
CAS Number: 10510-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98fb9f53cbe09539ebc3c0f2b70df8ab534153d8&vid=98fb9f53cbe09539ebc3c0f2b70df8ab534153d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
λmax (ethanol) 596 - 602 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR