4,4′-Azo-bis(4-cyanovaleric acid) ≥98%, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215041205 0215041225
IC15041205EA 100.11 USD
IC15041205 IC15041225
4,4′-Azo-bis(4-cyanovaleric acid) ≥98%, crystalline powder
4,4′-Azo-bis(4-cyanovaleric acid)

Soluble in water.(Lit.)

Formula: C₁₂H₁₆N₄O₄
MW: 280.284 g/mol
MDL Number: MFCD00002799
CAS Number: 2638-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR