4-Aminoantipyrine, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

0215034310
IC15034310EA 59.78 USD
IC15034310
4-Aminoantipyrine, yellow powder
4-Aminoantipyrine

4-Aminoantipyrine is a reagent for determination of chlorinated phenols, phenylglycosidase, and glucose in the presence of phenol and peroxidase.


Soluble in water, ethanol, benzene; sparingly soluble in ether.


Caution: Research Use Only (RUO).

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
MDL Number: MFCD00003145
CAS Number: 83-07-8
Merck Index: 13,00596

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 104 - 114 °C
UV/VIS: λmax (water) Record Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR