Agar (from Red Alga Rhodophyceae), off-white powder, Bacteriological Grade for bacteriology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Agar Agar

0215017890 0215017880 0215017883
IC15017890EA 506.22 USD
IC15017890 IC15017880 IC15017883
Agar (from Red Alga Rhodophyceae), off-white powder, Bacteriological Grade for bacteriology
Agar

Agar is a polysaccharide complex extracted by bleaching and hot water treatment of agarocytes from the red alga Rhodophyceae, and usually consisting of the genera Gelidium, Acanthopeltis, Ceramium, Pterocladia and Gracilaria. The algae are typically found in the Pacific and Indian Oceans and in the Sea of Japan. It is primarily composed of two different units: Agarose and Agaropectin; Agarose is a neutral gelling component which is composed of a linear polymer of alternating D-galactose and 3,6-anhydrogalactose units. Agaropectin is a non-gelling component which consists of D-1,3-glycosidically linked D-galactose units, some of which are sulfated at the 6th position.
Agar is specially purified for use in preparing solid culture media for microbiological and bacteriological applications.


  • Bacteriological Grade
  • Presentation: Off-white Powder
  • pH: 7.0 to 7.6 (1.5% aqueous solution) (Lit.)
  • Agar is strongly hydrophilic and can slowly absorb about 20 times its weight of cold water, swelling in the process. Hot water is usually necessary to solubilize agar. Insoluble in alcohols.
  • Storage temperature: Room Temperature


Total Ash ≤6.5% (dry weight basis); Acid Insoluble Ash ≤0.5%; Foreign Organic Matter ≤1.0%; Foreign Insoluble Matter ≤1.0%

Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Salmonella Negative
Loss on Drying ≤20%
Total Ash (dry weight basis) ≤6.5%
Acid Insoluble Ash ≤0.5%
Foreign Organic Matter ≤1.0%
Foreign Insoluble Matter ≤1.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR