Caffeine, anhydrous, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215011480 0215011490 0215011491
IC15011480EA 64.6 USD
IC15011480 IC15011490 IC15011491
Caffeine, anhydrous, powder
Caffeine
Formula: C₈H₁₀N₄O₂
MDL Number: MFCD00005758
CAS Number: 58-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21e44b791ffdd32d2de340516a29122483d41ce1&vid=ddad760108e519e7ed3a81124f4203755124401b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR