Ammonium chloride, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215010980
IC15010980EA 49.59 USD
IC15010980
Ammonium chloride, powder
Ammonium chloride

Soluble in water (100 mg/mL- clear, colorless solution). The presence of hydrochloric acid or sodium chloride decrease water solubility. Also soluble in methanol and ethanol.


Soluble in water (100 mg/mL- clear, colorless solution). The presence of hydrochloric acid or sodium chloride decrease water solubility. Also soluble in methanol and ethanol.

Formula: NH₄Cl
MDL Number: MFCD00011420
CAS Number: 12125-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR