Formamide, clear, colorless solution for molecular biology, deionized

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Carbamaldehyde

11FORMD002
IC11FORMD002EA 251.77 USD
IC11FORMD002
Formamide, clear, colorless solution for molecular biology, deionized
Formamide

Deionized Formamide is used in molecular biology and a denaturing agent for nucleic acids in gel electrophoresis orr hybridization experiments. In the latter case the role of formamide is to decrease the temperature necessary for the reassociation of complementary nucleic acids.


  • Grade : Molecular Biology Grade
  • Assay : > 99.5 %
  • Concentration : Neat
  • Key Applications : Used in Southern and Northern analyses, 50% or greater concentration for in situ hybridization techniques.
  • Application Areas : Nucleic Acid Electrophoresis; Nucleic Acid Sequencing
  • Product Type : Reagents
  • Lab Essential Type : Electrophoresis Biochemicals
  • Biochemical Category : Denaturing agents
  • UV/Visible Absorbance : A260 (0.5M, H2O) : < 0.05, A280 (0.5M, H2O) : < 0.02
  • Presentation : Clear colorless solution
  • Format : Solution
  • Conductivity : s100 µS at 25°C
  • Heavy Metals : Heavy metals (as Pb) : < 0.00005 %||Iron: < 0.00005 %
  • NOTES : Iron < 0.00005 %
  • Certifications : RNase, DNase, Protease free

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210
Melting Pt: 2
Flash Pt: 79
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
UN: 2810
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Assay >99.5%
Conductivity <100 µS/cm
A260 (0.5 M aq soln) <0.05
A280 (0.5 M aq soln) <0.02
Heavy Metals (as Pb) <0.00005%
Iron <0.00005%
Rnase Not Detected
Dnase Not Detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR