FastDNA™ Spin Kit for Soil, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Nucleic acid isolation, Mini-prep kit used to isolate bacterial, fungal, plant and animal genomic DNA from soil and other environmental samples.
- Pack Size: 50 preparations
- Harndess: Ceramic Spheres, Vickers 800; Silica Spheres, Knoop 515; Glass Bead, Knoop 515
- Specific Gravity: Ceramic Spheres, 4; Silica Spheres, 2.5; Glass Bead, 2.5
The FastDNA SPIN kt for Soil is designed to isolate bacterial, fungal, plant and animal genomic DNA from soil and other environmental samples. Soil is processed with a FastPrep® instrument and lysing matrix E tubes designed to efficiently lyse all microorganisms.
The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications. Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
Cited in over 6000 scientific publications, the FastDNA SPIN kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediment, sludge, compost, manure, rhizosphere or waste water. The FastDNA SPIN Kit for Soil contains lysing matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors.
Kit contents: 50×2.0 ml tubes of lysing matrix E, 60 ml sodium phosphate buffer, 8 ml MT buffer, 25 ml of PPS solution, 66 ml binding matrix, 50 spin modules, 50 catch tubes, 12 ml concentrated SEWS-M, 20 ml DES, 200 µL BBS gel loading dye.
Reagent: SEWS-M - pH range: 6.4 to 6.8.
Caution: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department. If working with biological, toxic, hazardous or infectious samples use hand protection (Latex, Nitrile or other protective approved material gloves), skin and body protection (Labcoat and long-sleeves), and eye protection (safety glasses with side-shields). Handle in accordance with good industrial hygiene and safety practices.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...