M9 Minimal Salts (Maniatis Formulation) Powder, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
M9 Minimal medium salts are suitable for the preparation of M9 Minimal Medium for use in propagation of
- Nonselective for Escherichia coli and Coliforms
- M9 medium can be used for maintaing positive selection pressure on plasmids
- Maintenance of F’-containing bacteria strains for M13 propagation
- Can be made as a sterile 5X concentrated solution and diluted to 1X before use
Media Preparation: Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in M9 Minimal Medium Salt formulations. Chosen reagents are combined in the proper proportions according to the formulation listed in the Certificate of Analysis and thoroughly blended for a uniform distribution.
coli and plasmid amplification.