LB Medium, Lennox Powder, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
113002122
IC113002122EA 135.87 USD
IC113002122
LB Medium, Lennox Powder, MP Biomedicals
Cell Culture Media
Luria-Bertani, Rich Medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
  • Proprietary media for enhanced cell yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
LB medium lennox powder is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. The growth characteristics are the same as L-broth agar powder. L-Broth supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.
Certifications: Molecular Biology Certified™.