EZ-Yeast™ Transformation Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
112100200
IC112100200EA 110.7 USD
IC112100200
EZ-Yeast™ Transformation Kit, MP Biomedicals
Nucleic Acid Reagents Transformation Reagents
The EZ-Yeast™ Transformation Kit is designed for high-throughput transformation with efficiencies up to 103 transformants per µg plasmid DNA.
  • Ideal where large number of transformations must be performed
  • Perfect for a quick analysis of potentially positive two-hybrid colonies
  • Does not require fully competent cells prior to transormation
The EZ-Yeast Transformation Kit is designed for high throughput transformations
Kits are for Cloning and Expression, Transformation, Yeast Cell Culture, Cell Viability Analysis, Microbiological Analysis.
It is ideal for cases where a large number of transformations must be performed, but only a few transformants are needed. Simply remove a 3-4 mm colony or 0.5 mL overnight culture to a new tube, add vector and carrier DNA, incubate for 30 minutes and plate on selective media.