Methyl laurate, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

0210526325
IC10526325EA 212.57 USD
IC10526325
Methyl laurate, liquid
Methyl laurate

water: insoluble (0.00759 g/L at 25 °C (practically))

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266
Melting Pt: 4.5
MDL Number: MFCD00008966
CAS Number: 111-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR