Monoolein, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 9-Octadecenoicacid(Z)-,2,3-dihydroxypropylester, 1-(cis-9-Octadecenoyl)-rac-glycerol, Glyceryl cis-9-octadecenate hygroscopic, DL-α-Monoolein, rac-Glycerol 1-monooleate, 1-Oleoyl-rac-glycerol, 1-Mono(cis-9-octadecenoyl)-rac-glycerol, 1-Monleoyl-rac-glycerol, Glyceryl monooleate

0210488880
IC10488880EA 94.41 USD
IC10488880
Monoolein, liquid
Monoolein

Glyceryl Monooleate is a surfactant that releases free glycerol and oleic acid upon hydrolysis.


Glyceryl Monooleate has been used in liquid crystal studies and research shows that in the presence of monoolein, the penetration of the drug cisplatin is doubled. Monoolein (1-Oleoyl-rac-glycerol) is used in the development of monoolein-based nanoparticulate liquid dispersions as possible vehicles for drug delivery.


Soluble in chloroform (50 mg/mL). Also soluble in hexane.


Glyceryl Monooleate is a surfactant that releases free glycerol and oleic acid upon hydrolysis.


Caution: Research Use Only (RUO).

Formula: C₂₁H₄₀O₄
MW: 356.547 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00042735
CAS Number: 111-03-5

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR