Peptone S from Soybean Meal, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0210480705
IC10480705EA 733.64 USD
IC10480705
Peptone S from Soybean Meal, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Peptone S is useful for incorporation into media used in hydrogen sulfide tests.
  • Digestion: Papain digest of soybean meal
Digestion: Papain digest of soybean meal
Peptone S is useful for incorporation into media used in hydrogen sulfide tests.