Yeast Hydrolysate Enzymatic, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210330401
IC10330401EA 70.83 USD
IC10330401
Yeast Hydrolysate Enzymatic, MP Biomedicals
Media for Microbiology Base Materials for Microbiology

Yeast Hydrolysate Enzymatic from Brewers Yeast


  • Source of soluble protein components, carbohydrates, and B Complex vitamins
  • Brown Powder
  • Soluble in water


Yeast Hydrolysate Enzymatic is an autolyzed yeast extract produced from 100% brewers yeast


Foreign activity - Salmonella: Negative
Notes
Protein (N x 6.25): 50.0-66.0%
Standard Plate Count (35°C): ≤50,000 CFU/g


It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR