Polysorbate 20, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210316880 0210316890 0210316891
IC10316880EA 46.71 USD
IC10316880 IC10316890 IC10316891
Polysorbate 20, liquid
Polysorbate 20

Polysorbate 20 is nonionic surfactant derived from sorbitan ester.


  • Refractive Index: n20/D 1.468(Lit.)
  • Viscosity: 400 cps at 25 °C.(Lit.)
  • Critical Micelle Concentration (CMC): 0.06mM


Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions, particularly in pharmaceutical applications. It has been used in pre-extraction of membranes to remove peripheral proteins. It has been used as a blocking agent on nitrocellulose.


Saponification No: 40-50


Polysorbate 20 is nonionic surfactant derived from sorbitan ester. Autoclaving of solutions is not recommended. Sterile filtering is suggested with a 0.22 micron filter. Tween® may need to be warmed to about 40 °C and alternated with portions of hot distilled water while being poured through the filter. As a blocking agent on nitrocellulose at a typical concentration of 0.05%. Used at concentration of 2% for extraction of membrane-bound proteins.


Caution: Research Use Only (RUO).

Formula: C₅₈H₁₁₄O₂₆
Boiling Pt: >100 °C (lit.)
Density: 1.095 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Specification Test Results

Identity Test A Passes
Identity Test B Passes
FTIR Conforms to Reference
Solubility (1g/20mL) Clear, Foamy Solution
Specific Gravity 1.08-1.113
Water Content ≤3%
Saponification Number 40-50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR