L(-)-Sorbose ≥98%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one

0210293990 0210293991
IC10293990EA 1013.3 USD
IC10293990 IC10293991
L(-)-Sorbose ≥98%, white crystalline powder
L(-)-Sorbose

Sorbose is a ketose belonging to the group of sugars known as monosaccharides. It has a sweetness that is equivalent to sucrose.The commercial production of vitamin C (ascorbic acid) often begins with sorbose


L-Sorbose is used as a starting reagent for the commercial biosynthesis of ascorbic acid (vitamin C).


Freely soluble in water (50 mg/ml-clear, colorless solution); almost insoluble in alcohol.


Sorbose is a ketose belonging to the group of sugars known as monosaccharides. It has a sweetness that is equivalent to sucrose.The commercial production of vitamin C (ascorbic acid) often begins with sorbose

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 380
Storage Temperature: Ambient
MDL Number: MFCD00151097
CAS Number: 87-79-6
Merck Index: 12,08874

Order Now

Specification Test Results

Identity Test Passes
Purity (HPLC) ≥98%
FTIR Conforms to Standard
Melting Point 159 - 175 °C
Optical Rotation:(c=2,water) −38° to −48°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR