Sodium selenite ∼98%, powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210292825
IC10292825EA 109.82 USD
IC10292825
Sodium selenite ∼98%, powder cell culture reagent
Sodium selenite

Sodium selenite is commonly used as a source of selenium in biological research, where selenium is an essential trace element that is normally provided by serum. Selenium is present in selenoproteins such as glutathione peroxidase and thioredoxin reductase, which contain the selenium analog of cysteine, selenocysteine. In particular, glutathione peroxidase has a role in detoxification in vivo as a scavenger of peroxides. Sodium selenite is included in various medium supplements for use in cell culture. Sodium selenite has been utilized in studies of cell proliferation and cancer research. It has been used to alter gene expression in HepG2 cells as analyzed by cDNA microarrays. Sodium selenite can inhibit zinc finger protein/DNA interactions. Sodium selenite has been shown to alter mitochondrial membrane potentials and thus potentially contribute to apoptosis.


More potent than selenomethionine in reducing carcinogenesis.


Selenium Content ≥44.0%


Soluble in water.


Sodium selenite is commonly used as a source of selenium in biological research, where selenium is an essential trace element that is normally provided by serum. Selenium is present in selenoproteins such as glutathione peroxidase and thioredoxin reductase, which contain the selenium analog of cysteine, selenocysteine. In particular, glutathione peroxidase has a role in detoxification in vivo as a scavenger of peroxides. Sodium selenite is included in various medium supplements for use in cell culture. Sodium selenite has been utilized in studies of cell proliferation and cancer research. It has been used to alter gene expression in HepG2 cells as analyzed by cDNA microarrays. Sodium selenite can inhibit zinc finger protein/DNA interactions. Sodium selenite has been shown to alter mitochondrial membrane potentials and thus potentially contribute to apoptosis.

Formula: Na₂SeO₃
MW: 172.94 g/mol
MDL Number: MFCD00003489
CAS Number: 10102-18-8
UN: 2630
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR