Nicotinamide, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210244780 0210244791
IC10244780EA 30.14 USD
IC10244780 IC10244791
Nicotinamide, Powder
Nicotinamide

Soluble in water (50 mg/mL).


Readily Carbonizable Substances: Conforms to USP


Niacinamide is used as a cofactor for certain enzymes.

Formula: C₆H₆N₂O
MDL Number: MFCD00006395
CAS Number: 98-92-0

Order Now

Specification Test Results

Synonyms Nicotinic acid amide; Nicotinamide; Pyridine-3-carboxylic acid amide; Vitamin B3; Vitamin PP
Form White Powder
Storage Store at Room Temperature(15-30 °C).
Assay 98.5 - 101.5% (dried basis)
Heavy Metals ≤0.003%
Loss on Drying ≤0.5%
Residue on Ignition ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR