D-(-)-Mannitol ≥96%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

02102248.5 0210224801 0210224805
IC102248.5EA 55.97 USD
IC102248.5 IC10224801 IC10224805
D-(-)-Mannitol ≥96%, powder
D(-)-Mannitol

Mannitol is carbohydrate found in plants and plant exudates.


A sugar alcohol sweet tastant. Used in sweetness inhibition studies.


Soluble in water (1 g/5,5 ml), alcohol (1 g/83 ml), glycerol (1 g/18 ml); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.


Mannitol is carbohydrate found in plants and plant exudates.

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Melting Pt: 163…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Manna sugar; Mannite
Form Powder
Storage Store at Room Temperature (15-30 °C).
Loss on Drying ≤1.0%
Optical Rotation +130 to +150°(c=1, ammonium molybdate, sulfuric acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR