Maleic acid, white crystalline powder, Technical Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210223201
IC10223201EA 191.83 USD
IC10223201
Maleic acid, white crystalline powder, Technical Grade
Maleic acid

Maleic acid (MA) is a dicarboxylic acid that undergoes esterification with ethanol in the presence of cation-exchange resin catalysts to form diethyl maleate. Its crystalline structure has been analyzed. Reaction kinetics and mechanism of the isomerization of maleic acid to fumaric acid in the presence of ceric and bromide ions has been investigated. The reaction mechanism and reactivity ratios for the radical copolymerization of maleic acid with styrene have been determined.


Soluble in Water. (1% aq soln, 333 mg/mL).

Formula: C₄H₄O₄
MDL Number: MFCD00063177
CAS Number: 110-16-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to standard
Moisture Content ≤5%
Melting Point 130 - 145°C
Solubility (1% aq.) Clear and colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR