Lecithin, viscous liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Lecithin is a phosphatidylcholine of soybean origin.
Lecithin is a good source of choline, which may help with brain function and growth. Used as an emulsifier and solubilizer for drugs in an aqueous solution, also as a vehicle for liposomes.
Acid Value: ≤32
Soluble in about 12 parts cold, absolute alcohol; soluble in chloroform, ether, petr ether, in mineral oils and fatty acids; sparingly soluble in benzene. Insoluble in acetone.
Lecithin is a phosphatidylcholine of soybean origin.
MDL Number:
MFCD00147406 CAS Number: 8002-43-5 |
Specification Test Results
Synonyms | L-a-Phosphatidylcholine; 1,2-Diacyl-sn-glycero-3-phosphocholine; L-alpha-Lecithin; 3-sn-Phsophatidylcholine; L-alpha-Phosphatidylcholine |
Form | Viscous Golden Brown Liquid |
Storage | Store at -0 °C. |
Insoluble Matter | Acetone Insolubles: ≥62%; Hextane Insolubles: ≤0.1% |
Moisture Content | Moisture: ≤1% |
Source | Soybean |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...