4-Hydroxy-2-quinolinecarboxylic acid, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 4-Hydroxyquinoline-2-carboxylic Acid, Transtorine, Empirical Formula, Kynurenic acid, Quinurenic acid, KYNA, Kynuronic acid, Kinurenic acid

02102124.2
IC102124.2EA 35.04 USD
IC102124.2
4-Hydroxy-2-quinolinecarboxylic acid, powder
4-Hydroxy-2-quinolinecarboxylic acid

Kynurenic acid is a non-selective antagonist at NMDA and AMPA/kainate receptors.


  • Blocks kainic acid neurotoxicity
  • Blocks nicotinic acetycholine receptors


Soluble in water, hot alcohol and 1.0 M NaOH (50 mg/ml-clear, pale yellow solution); insoluble in ether


Kynurenic acid is a non-selective antagonist at NMDA and AMPA/kainate receptors.


Caution: Research Use Only (RUO).

Formula: C₁₀H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00149476
CAS Number: 492-27-3

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR