Estradiol ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

02101656.2 0210165601 0210165605 0210165680
IC101656.2EA 77.9 USD
IC101656.2 IC10165601 IC10165605 IC10165680
Estradiol ≥98%, white powder
Estradiol

β-Estradiol is used to study cell differentiation and transformations (tumorigenicity). The pharmacology and HPLC determination of β-Estradiol in plasma and urine have been reported.


β-Estradiol is almost insoluble in water. Freely soluble in alcohol; soluble in acetone, dioxane, other organic solvents; solutions of fixed alkali hydroxides; sparingly soluble in vegetable oils; soluble in methanol : CHCl3 (1:1). Dissolve 50 mg/mL in ethanol with heating and observed a clear, colorless solution. It is also soluble in acetone and dioxane but only sparingly soluble in vegetable oils.


β-Estradiol is used to study cell differentiation and transformations (tumorigenicity). The pharmacology and HPLC determination of β-Estradiol in plasma and urine have been reported.


Caution: Research Use Only (RUO).

Formula: C₁₈H₂₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00003693
CAS Number: 50-28-2

Order Now

Specification Test Results

Identity Test Passes
Assay ≥98%
FTIR Conforms to Standard
Melting Point 173 - 179 °C
Optical Rotation +76° to +83° (c = 1% in dioxane at 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR