6-Aminocaproic acid, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

0210164590 0210164591
IC10164590EA 325.01 USD
IC10164590 IC10164591 IC0210164594
6-Aminocaproic acid, white powder
6-Aminocaproic acid

ε-Amino-n-Caproic Acid is a plasmin inhibitor. It inhibits carboxypeptidase B. It promotes rapid dissociation of plasmin by inhibiting the activation of plasminogen. As an inhibitor of plasmin it is has been utilized in the clotting buffer for fibrinogen assays.


EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carbo­xy­peptidase, plasmin, and plasminogen activator. Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.


Soluble in water (50 mg/ml); sparingly soluble in alcohol and methanol; practically insoluble in ether; insoluble in ethanol.


ε-Amino-n-Caproic Acid is a plasmin inhibitor. It inhibits carboxypeptidase B. It promotes rapid dissociation of plasmin by inhibiting the activation of plasminogen. As an inhibitor of plasmin it is has been utilized in the clotting buffer for fibrinogen assays.


Caution: Research Use Only (RUO).

Formula: C₆H₁₃NO₂
Melting Pt: 206,7 °C
Density: 1,48 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 205 - 215 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR