tert-Butyl-4-methoxyphenol (mixture of isomers) ∼98.5%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0210115980 0210115990
IC10115980EA 54.46 USD
IC10115980 IC10115990
tert-Butyl-4-methoxyphenol (mixture of isomers) ∼98.5%, white powder
tert-Butyl-4-methoxyphenol (mixture of isomers)

Butylated Hydroxyanisole is an antioxidant. It is a mixture of 2- and 3-tertiary-butyl-4-hydroxyanisole.


Freely soluble in ethanol, methanol (50 mg/mL - clear and colorless) and in propylene glycol. Soluble in fats and oils. Insoluble in water


Butylated Hydroxyanisole is an antioxidant. It is a mixture of 2- and 3-tertiary-butyl-4-hydroxyanisole.


Caution: Research Use Only (RUO).

Formula: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD01779059
CAS Number: 25013-16-5

Order Now

Specification Test Results

Identity Test Passes
Purity ~98.5%
FTIR Conforms to Reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR