(±)-α-Lipoic acid ≥98%, Powder

Supplier: 0000012207

0210113801 0210113805 0210113825 0210113850
IC10113801EA 42.1 USD
IC10113801 IC10113805 IC10113825 IC10113850
(±)-α-Lipoic acid ≥98%, Powder
(±)-α-Lipoic acid

DL-Thioctic Acid is a hydrogen transferrin cofactor.


Antioxidant and coenzyme needed for the activity of enzyme complexes such as those of pyruvate dehydrogenase and glycine decarboxylase. Exogenous thioctic acid is reduced intracellularly by two or more enzymes. The reduced form then influences a number of cell processes by direct radical scavenging, recycling of other antioxidants, increasing glutathione synthesis, and modulating transcription factor activity particularly that of NF-κB. Decreases the phagocytosis of myelin by macrophages.


Soluble in fat solvents, methanol (50 mg/mL-clear pale yellow solution), ethanol(50 mg/mL-clear solution); practically insoluble in water.


DL-Thioctic Acid is a hydrogen transferrin cofactor.

Formula: C₈H₁₄O₂S₂
MDL Number: MFCD00005474
CAS Number: 1077-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR