Bicine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

0210100525 0210100580 0210100583 0210100591
IC10100525EA 65.62 USD
IC10100525 IC10100580 IC10100583 IC10100591
Bicine, white powder
Bicine

Bicine is an organic compound used as a buffering agent. It is a zwitterionic buffer and one of the Good's buffers . It is prepared by the reaction ofglycine with ethylene oxide, followed by hydrolysis of the resultant lactone. It is a recommended buffer for low temperature biochemical work. Used for the preparation of stable substrate solution for serum guanase determination.


Bicine is used in a thin layer ion exchange chromatography method for protein resolution.It is used in peptide and protein crystallization.Bicine is used in a kinetic study of a quaternary transition-state analogue complex of creatine kinase and in multiphasic buffer system for SDS-PAGE of proteins.


Soluble in water (10 mg/ml - clear, colourless solution; a saturated solution in water is 1,1 M )

Formula: C₆H₁₃NO₄
Melting Pt: 190 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Reference
pH (1% aq soln) 4.0 - 6.0
OD260 nm (10% aq soln) 0.100
OD280 nm (10% aq soln) 0.100


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR