6-Benzylaminopurine, Powder

Supplier: 0000012207
Warning

02100912.5 0210091205 0210091225
IC100912.5EA 50.72 USD
IC100912.5 IC10091205 IC10091225
6-Benzylaminopurine, Powder
6-Benzylaminopurine

6-Benzylaminopurine (BAP) is a plant cell division factor (a plant growth factor).


6-Benzylaminopurine, benzyl adenine (BAP) is a synthetic cytokinin which together with auxins elicits plant growth and development responses. BAP is a widely use cytokinin supplement to plant growth media such as Murashige and Skoog medium, Gamborg′s medium, and Chu′s N6 medium. Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables.


BAP is an inhibitor of respiratory kinase in plants, and increases post-harvest life of green vegetables.


E (270) = ~19,054 (methanol)(Lit.)


Soluble in glacial Acetic Acid (50 mg/mL). Soluble in concentrated ammonium hydroxide, 0.1 N Sodium hydroxide (10 mg/mL - slightly hazy and yellowish solution), methanol, or dioxane.


6-Benzylaminopurine is a plant cell division factor (a plant growth factor).


Caution: Research Use Only (RUO).

Formula: C₁₂H₁₁N₅
Storage Temperature: Ambient
MDL Number: MFCD00005572
CAS Number: 1214-39-7

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 226 - 236 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR