D-(-)-Arabinose ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

0210069925 0210069980
IC10069925EA 160.14 USD
IC10069925 IC10069980
D-(-)-Arabinose ≥99%, powder
D(-)-Arabinose

D-( -)-Arabinose is a D-ribose analogue present in sugar polymers such as the arabinogalatans of mycobacterial cell walls and plant tissues.


D-( -)-Arabinose is a substrate used to identify, differentiate and characterize arabinose isomerase(s) that have commercial value in the production of tagatose, a low calorie sweetener.


Soluble in water (50 mg/mL) giving clear, colorless to faint yellow solution.


D-Arabinose is a reducing sugar. It is a pentose analog of D-ribose that is a constituent of mycobacterial cell wall arabinogalactans. It is also a substrate for D-erythroascorbic acid synthesis in yeast.

Formula: C₅H₁₀O₅
MDL Number: MFCD00135608
CAS Number: 10323-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR