Dithiothreitol (DTT, Cleland's reagent) ≥99.0%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

02100597.2 0210059701 0210059705 0210059710 0210059725 0210059750 0210059780
IC100597.2EA 50.8 USD
IC100597.2 IC10059701 IC10059705 IC10059710 IC10059725 IC10059750 IC10059780
Dithiothreitol (DTT, Cleland's reagent) ≥99.0%, white powder
Dithiothreitol (DTT, Cleland's reagent)

DL-Dithiothreitol is also known as Clelands reagent; Protective agent for sulfhydryl groups (-SH). Quantitatively reduces disulfides (-S-S- to -SH). In this reaction the DTT is oxidized to the cyclic disulfide which ensures the reduction of other disulfides in solution. Disulfide reduction occurs quickly at pH 8.

DTT has been used:


  • as one of the reactants in the reduction and alkylation of αs1-Casein, the major allergen of cow′s milk.
  • as a component of medium for the demembranation and reactivation of spermatozoa.
  • to maintain stability of the enzyme as thiol effectively protects the active sites of the biocatalyst.
  • as a reducing agent to test the specificity of the reaction of N-Ethylmaleimide with sulfhydryl groups.


Dithiothreitol is useful for stabilizing sulfhydryl containing enzymes. Effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).


Dithiothreitol is useful for stabilizing sulfhydryl containing enzymes. Effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).It is used in preparation of carboxymethylated rPA (IAA-rPA)(Alexander Tischer(2010), l-Arginine hydrochloride increases the solubility of folded and unfolded recombinant plasminogen activator rPA, Protein science,19(9), 1783-1795 ).


Soluble in water (50 mg/mL - clear and colorless solution); soluble in ethanol, acetone, ethylate, chloroform and ether.


Caution: Research Use Only (RUO).

Formula: C₄H₁₀O₂S₂
Boiling Pt: ∼125 °C (1013 hPa)
Melting Pt: 40…50 °C (lit.)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 27565-41-9
EINECS: 222-468-7
UN: 3335
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity (Titration) ≥99.0%
Melting Point 40 - 50 °C
FTIR Conforms to Standard
OD Readings For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR