Penicillin G potassium salt, Powder

Supplier: 0000012207

0210054310 0210054380 0210054325
IC10054310EA 77.7 USD
IC10054310 IC10054380 75875-628
Penicillin G potassium salt, Powder
Penicillin G potassium salt

Penicillin G is a narrow spectrum antibiotic


Penicillin G is active against gram-positive and gram-negative aerobic and anaerobic bacteria. Penicillin G inhibits cell wall synthesis by binding to penicillin binding proteins (PBPs) which inhibits peptidoglycan chain cross-lining. Penicillin G is stable against hydrolysis by β-lactamases, such as penicillinases and cephalosporinases.


Antimicrobial spectrum: This product is active against gram-positive and gram-negative bacteria.


Mode of Action: Penicillin G acts by inhibiting cell wall synthesis through binding to penicillin binding proteins (PBPs), inhibiting peptidoglycan chain cross-linking..

Formula: C₁₆H₁₇KN₂O₄S
Melting Pt: 214…217 °C (lit.)
MDL Number: MFCD00036193
CAS Number: 113-98-4

Order Now

Specification Test Results

Synonyms Benzylpenicillin potassium salt
Form White Powder
Storage Store at +4 °C.
pH 5 - 7.5 (3% aq soln)
Potency 1500-1700 u/mg
Solubility Soluble in water (100mg/mL) clear,colorless to light yellow.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR