Iodoacetamide, tan powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0210035105 0210035110 0210035125 0210035191
IC10035105EA 78.03 USD
IC10035105 IC10035110 IC10035125 IC10035191
Iodoacetamide, tan powder
Iodoacetamide

Iodoacetamide is a thiol reagent; alkylating reagent for cysteine and histidine residues in proteins. In the alkylation reaction, it reacts with histidine (such as in RNase), methionine and sulfhydryl groups of many proteins.


Iodoacetamide can react with low molecular weight thiol compounds such as mercaptoethanol and glutathione. The reaction with glutathione is utilized in a titration method for the determination of sulfhydryl groups in different compounds. The alkylation reaction forms stable protein derivatives which will remain intact during further study of the protein.


Iodoacetamide is useful in peptide sequencing and as an irreversible enzyme inhibitor. Some enzymes iodoacetamide will inhibit include (but is not limited to): Alcohol dehydrogenase, Alkaline phosphatase (calf intestinal), beta-Amylase, Cathepsin B, Galactose oxidase, Heme oxygenase, L-Lysine decarboxylase. It will irreversibly inhibit the dephosphorylation of both epidermal growth factor receptor and platelet-derived growth factor receptor by alkylation at the active center of one or several protein tyrosine phosphatases in cell lines. Iodoacetamide will also irreversibly inhibit many cysteine proteases. Alkylating reagent for cysteine residues in peptide sequencing. By virtue of reaction with cysteine, it is an irreversible inhibitor of enzymes with cysteine at the active site. It reacts much more slowly with histidine residues, but that activity is responsible for inhibition of ribonuclease.


Soluble in water (recommended stock solution is 10-100 mM). All solutions should be prepared fresh for each use since solutions are very light sensitive.


Iodoacetamide is a thiol reagent; alkylating reagent for cysteine and histidine residues in proteins. In the alkylation reaction, it reacts with histidine (such as in RNase), methionine and sulfhydryl groups of many proteins. Iodoacetamide can react with low molecular weight thiol compounds such as mercaptoethanol and glutathione. The reaction with glutathione is utilized in a titration method for the determination of sulfhydryl groups in different compounds. The alkylation reaction forms stable protein derivatives which will remain intact during further study of the protein.

Formula: ICH₂CONH₂
MDL Number: MFCD00008028
CAS Number: 144-48-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cea6c7bd88d1b733b033eb1caa008dee7cdcfdac&vid=cea6c7bd88d1b733b033eb1caa008dee7cdcfdac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR