Adonitol, white powder
Supplier: 0000012207
Adonitol (ribitol), a pentose alcohol, is metabolised to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.
Adonitol is a crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution.
Soluble in water (100 mg/ml - clear, colorless solution)
Adonitol is a crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution.
Caution: Research Use Only (RUO).
Formula:
HOH₂C(CH(OH))₃CH₂OH |
MDL Number:
MFCD00064291 CAS Number: 488-81-3 |
|
Specification Test Results
Identity Test | Passes |
Melting Point | 98 - 106 °C |
FTIR | Conforms to Standard |
Moisture Content |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...