5-Bromo-2′-deoxycytidine hydrate, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
|
0210016680
0210016691
IC10016680EA
57.6
USD
IC10016680
IC10016691
5-Bromo-2′-deoxycytidine hydrate, white powder
5-Bromo-2′-deoxycytidine hydrate
5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP.
Soluble in water, NH₄OH (50 mg /ml).
Formula:
C₉H₁₂BrN₃O₄·×H₂O MW: 306.116 g/mol (anhy) Storage Temperature: Freezer |
CAS Number:
1022-79-3 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
UV/VIS: λmax (water) | 286 - 289 nm |
Moisture | Report Result |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...