Microplate Sealing Tape, MP Biomedicals
Supplier: MP Biomedicals
097742000
IC097742000EA 111.74 USD
IC097742000
Microplate Sealing Tape, MP Biomedicals
Seals Microplate Seals
Acetate plate sealers are not gas permeable and not autoclavable.
  • This microplate sealing tape is easy to apply and remove from microplates
  • Microplate sealing tape is suitable for all microplates for both short and long term storage
  • Provides a tight seal to minimize evaporation and condensation
  • 60 m per roll
Caution: Store at room temperature.