Acetate Plate Sealer, Nonsterile, MP Biomedicals
Supplier: MP Biomedicals
0976401C2
IC0976401C2BX 673.85 USD
IC0976401C2
Acetate Plate Sealer, Nonsterile, MP Biomedicals
Seals Microplate Seals
Acetate plate sealer is easy to apply and remove from microplates. In addition, this microplate sealing tape is suitable for all microplates for both short and long term storage, providing a tight seal to minimize evaporation and condensation.
  • Easy application and removal for all microplates for both short and long term storage
  • Clear film with adhesive on one side
  • Suitable for use between –16 and 38 °C
  • Tape is transparent and is not pierceable
  • Detection method: FTIR
Non-gas permeable and not autoclavable. Use for enzyme-linked immunoassays (ELISA) and Polymerase chain reaction (PCR).