Multitest Slide, 12-Well, MP Biomedicals
Supplier: MP Biomedicals
096041205
IC096041205BX 219.36 USD
IC096041205
Multitest Slide, 12-Well, MP Biomedicals
Slides Chamber Slides
Uncoated areas have a total wettable surface so that liquid spreads evenly over the area of each well. For cell attachment and growth, the slides require only a simple washing in deionized water or detergent followed by sterilization.
  • Applications include immunofluorescence, cell culture, slide agglutination tests, and histology
  • Features blue coating
  • Fragile glass
  • Well diameter is 7 mm
  • Well orientation is 2×6
A four channel pipette can be used to transfer samples directly from the microplate to the slide.
Caution: The Multitest Slide is not a reusable slide. Soaking and washing after use can cause the PTFE coating to lift off due to previous reactions of organic solvents and fixatives.