Human Type AB Serum, MP Biomedical
Supplier: MP Biomedicals
092930149
IC092930149EA 349.29 USD
IC092930149
Human Type AB Serum, MP Biomedical
Sera (Serum)
Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.
  • Collected from Type AB human blood
  • Liquid media
  • Storage at —20 °C
It has been proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.