EDTA disodium salt solution 0,2% (w/v) (synthetic)

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

092820549
IC092820549EA 26.71 USD
IC092820549
EDTA disodium salt solution 0,2% (w/v) (synthetic)
EDTA disodium salt

EDTA solution for use as a gentle non-enzymatic cell dissociation reagent.
Solution in Phosphate Buffered Saline without Calcium and Magnesium


  • Clear, colorless
  • pH: 4.0 - 6.0
  • Foreign activity: Mycoplasma: Negative
  • Osmolality: 240 - 450 mM/kg

Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
Merck Index: 12,03556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR