Alsever's Solution, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
092801154
IC092801154EA
80.79
USD
IC092801154
Alsever's Solution, MP Biomedicals
Cell Culture Media
Alsever's Solution is routinely used as an anti-coagulant/blood preservative, which permits the storage of whole blood at refrigerator temperatures for approximately 10 weeks.
- Isotonic, balanced salt solution
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)