Sodium pyruvate 11.00mg/ml 100 mM, sterile

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt, alpha-Ketopropionic acid sodium salt

091682049
IC091682049EA 22.1 USD
IC091682049
Sodium pyruvate 11.00mg/ml 100 mM, sterile
Sodium pyruvate

Sodium pyruvate is used by cells as an easily accessible carbohydrate source. Additionally, it is involved with amino acid metabolism and initiates the Kreb′s cycle. The 100 mM solution should be diluted 1:100 for most cell culture. The use of sodium pyruvate in Wallen fermentation medium to enhance the conversion of oleic acid to 10-ketostearic acid by Bacillus sphaericus has been described. A protocol that uses sodium pyruvate to establish stably transfected human B cell lines has been published. It improves coliform recovery when present in culture medium.


  • Application: Primary Cell Culture
  • pH: 6.8 (Lit.)
  • Osmolality: 430 - 500 mOsm/kg H2O


In muscle, pyruvic acid (derived from glycogen) is reduced to lactic acid during exertion, which is reoxidized and partially retransformed to glycogen during rest. Involved in a metabolic regulatory pathway activated by mitochondrial oxidants. Pyruvate is involved in respiratory regulation in plants by interacting with alternative oxidase at a conserved cysteine residue. May help prevent hydrogen peroxide mediated cell death. Pyruvate, the anion of pyruvic acid, is the end product of the glycolysis pathway, whereby glucose is converted to pyruvate with the production of ATP.

Formula: CH₃COCOONa
MW: 110.04 g/mol
Density: ~1.267 g/mL (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002586
CAS Number: 113-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR