Trace Element Solution 2, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
091676649
IC091676649EA 121.61 USD
IC091676649
Trace Element Solution 2, MP Biomedicals
Cell Culture Media
MP Biomedicals offers a 1000X sterile filtered solution for use with serum-free media.
  • To be added aseptically to media as a supplement
  • Same composition as Cleveland’s trace element I
Prior to use of media, it may be necessary to adjust pH and sterile filter (optional, reduces contamination).