Trace Element Solution 1, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
091676549
IC091676549EA 203.18 USD
IC091676549
Trace Element Solution 1, MP Biomedicals
Cell Culture Media
MP Biomedicals offers a 1000X sterile filtered solution for use with serum-free media.
  • Includes selenium, copper, zinc, and iron, as well as ferric citrate for greater stability