Washed, Preserved Sheep Red Blood Cells (10% Suspension), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0855876
IC0855876EA 276.57 USD
IC0855876
Washed, Preserved Sheep Red Blood Cells (10% Suspension), MP Biomedicals
Cells
Product is 20 mL of a 10% suspension of sheep red blood cells.
Sheep Red Blood Cells, 10% is suitable for immunoassays (rosetting), hemolytic and hemagglutination assays and for assays of heme inhibition.
Product reacts in an agglutination assay with anti-sheep red blood cells. The hematocrit is within the acceptable range.
Red blood cells collected into Alsever's solution (Dextrose, sodium chloride, sodium citrate), washed with phosphate buffered saline and preserved with borate buffer, sodium acetate.
Suspension of sheep red blood cells collected into Alsever's solution, washed with phosphate buffered saline and preserved with borate buffer, sodium acetate and 0.1% thimerosal