Anti-Seminal Fluid Rabbit Polyclonal Antibody

Supplier: MP Biomedicals
0855137
IC0855137EA 338.27 USD
IC0855137
Anti-Seminal Fluid Rabbit Polyclonal Antibody
Antibodies
Product is the lyophilized powder of rabbit antiserum to human serum and buffer salts.
The antiserum is prepared by hyperimmunizing rabbits with human serum. The antiserum is delipidated and dialyzed into 0.02M
sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22
µm filter, vialed and lyophilized.

Type: Primary 
Antigen: Seminal Fluid
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR