Cholesterol, water soluble

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

0219934230
IC0219934230EA 142.64 USD
IC0219934230
Cholesterol, water soluble
Cholesterol

Cholesterol is often used in cell culture applications.


Product is approximately 40 mg of cholesterol per gram containing a balance of methyl-β-cyclodextrin.

Formula: C₂₇H₄₆O
MW: 386.66 g/mol
MDL Number: MFCD00003646
CAS Number: 57-88-5

Order Now

Specification Test Results

Physical Appearance White Crystalline Powder
Solubility Soluble in water (300 mg/mL- Clear to Slightly Hazy solution), alcohol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR