Dimethyl sulfoxide ≥99% cell culture reagent

Supplier: 0000012207
Warning

0219605525 0219605580 0219605590 0219605591
IC0219605525EA 61.54 USD
IC0219605525 IC0219605580 IC0219605590 IC0219605591
Dimethyl sulfoxide ≥99% cell culture reagent
Dimethyl sulphoxide

Dimethyl Sulfoxide is ideal for use as a cryoprotectant. Also used in chemical reactions, in polymerase chain reactions (PCR) as a PCR cosolvent to help improve yields, especially in long PCR, and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal-induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells (ESC), and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum (FBS). Used to enhance dermal absorption of many chemicals. A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers. Used in antifreeze or hydraulic fluids. A cryopreservative for cell cultures. Used in the oxidation of thiols and disulfides to sulfonic acids.

Dimethyl sulfoxide (DMSO) is a highly polar aprotic organic reagent that has exceptional solvent properties for organic and inorganic chemicals

Physical Apppearance: Liquid
Viscosity: 1.1 centipoises (27 °C)
Vapor Density: 2.7 (vs air)
Vapor Pressure: 0.55 hPa at 20 °C, 0.42 mmHg (20 °C)
Refractive Index: n20/D 1.479(lit.)
Solubility: Soluble in water (1 mL DMSO + 1 mL H2O), methanol, acetone, ether, benzene, chloroform

Formula: (CH₃)₂SO
Boiling Pt: 189 °C (lit.)
Melting Pt: 16…19 °C (lit.)
Density: 1.104 g/cm³ (20 °C)
Flash Pt: 87 °C (188.6 °F) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cee06e48a98bf61d1893b8a6c3e1d17916df0d2&vid=4a96e98089ba441a0c4c9fb238031621a4562d00&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR